Assay Details
Target Gene Details
Entrez Gene ID: | 110460 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 2 |
Gene Aliases: |
AW742799, Tcp-1x, Tcp1-rs1 |
Location: |
Chr.17:12943042-12960725 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acat2 | NM_009338.3 | NP_033364.2 | ||
AK037249.1 | BAC29776.1 | |||
AK151751.1 | ||||
AK166813.1 | BAE39040.1 | |||
AK167234.1 | BAE39358.1 | |||
BC004823.1 | ||||
BC012496.1 |
More Information
Set Membership: |
Intragenic Non-exonic |