Assay Details
Target Gene Details
Entrez Gene ID: | 240084 |
Gene Name: | coiled-coil alpha-helical rod protein 1 |
Gene Aliases: |
Hcr |
Location: |
Chr.17:35517028-35531015 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cchcr1 | NM_146248.2 | 3 | 327 | NP_666360.2 |
AK159780.1 | 3 | 254 | BAE35364.1 | |
BC031416.1 | 4 | 416 | AAH31416.1 | |
BM947393.1 | 3 | 239 | ||
BY141405.1 |