Assay Details
Target Gene Details
Entrez Gene ID: | 245847 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
5730457F11Rik |
Location: |
Chr.17:24155833-24163733 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amdhd2 | NM_172935.4 | NP_766523.2 | ||
AK077584.1 | BAC36877.1 | |||
AK090009.1 | BAC41042.1 | |||
AK154209.1 | ||||
AK159860.1 | BAE35434.1 | |||
AK168432.1 | BAE40339.1 | |||
AK180149.1 | ||||
AK180231.1 | ||||
AK185305.1 | ||||
AK205862.1 | ||||
AK215432.1 | ||||
AK219479.1 | ||||
BC037005.1 | AAH37005.1 |
More Information
Set Membership: |
Intragenic Non-exonic |