Assay Details
Target Gene Details
Entrez Gene ID: | 53817 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B |
Gene Aliases: |
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh |
Location: |
Chr.17:35241746-35253707 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx39b | NM_001252457.1 | 5 | 706 | NP_001239386.1 |
NM_019693.3 | 5 | 794 | NP_062667.1 | |
AF118128.1 | 4 | 560 | AAD13115.1 | |
AK051034.1 | 5 | 727 | BAC34505.1 | |
AK088867.1 | 5 | 712 | BAC40624.1 | |
AK171507.1 | 5 | 736 | BAE42496.1 | |
AK179863.1 | 3 | 331 | ||
AK194985.1 | 1 | 111 | ||
AK195134.1 | 1 | 111 | ||
AK217230.1 | 1 | 81 | ||
AY255786.1 | 4 | 686 | AAP91685.1 | |
AY255787.1 | 4 | 686 | AAP91686.1 | |
BC011067.1 | 5 | 702 | AAH11067.1 | |
BC024859.1 | 4 | 1930 | AAH24859.1 |