Assay Details
Target Gene Details
Entrez Gene ID: | 26462 |
Gene Name: | thioredoxin reductase 2 |
Gene Aliases: |
AA118373, ESTM573010, TGR, Tr3, Trxr2, Trxrd2 |
Location: |
Chr.16:18426417-18479073 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Txnrd2 | NM_013711.3 | NP_038739.2 | ||
AB027566.1 | BAA86986.2 | |||
AF136399.1 | AAF03359.1 | |||
AF171053.1 | AAD51323.1 | |||
AF412308.1 | AAQ03230.1 | |||
AK008004.1 | BAB25403.1 | |||
AK075828.1 | ||||
BC013688.1 | AAH13688.1 | |||
BC052157.1 |
More Information
Set Membership: |
Intragenic Non-exonic |