Assay Details
Target Gene Details
Entrez Gene ID: | 224139 |
Gene Name: | golgi autoantigen, golgin subfamily b, macrogolgin 1 |
Gene Aliases: |
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151 |
Location: |
Chr.16:36875173-36933085 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golgb1 | NM_030035.1 | NP_084311.1 | ||
AK081762.1 | ||||
AK220188.1 | BAD90373.1 | |||
BC040461.1 | AAH40461.1 | |||
BC042759.1 | AAH42759.1 | |||
BC060254.1 | AAH60254.1 | |||
BC145221.1 | ||||
BC150731.1 |