Assay Details
Target Gene Details
Entrez Gene ID: | 27416 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 5 |
Gene Aliases: |
2900011L11Rik, AI132311, Abcc5a, Abcc5b, Mrp5 |
Location: |
Chr.16:20331303-20426394 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc5 | NM_013790.2 | 23 | 3519 | NP_038818.2 |
AB019003.1 | 23 | 3470 | BAA76609.1 | |
BC090629.1 | 23 | 3476 | AAH90629.1 |
More Information
Set Membership: |
Intragenic Exonic |