Assay Details
Target Gene Details
Entrez Gene ID: | 74374 |
Gene Name: | C-type lectin domain family 16, member A |
Gene Aliases: |
4932416N17Rik, curt, mKIAA0350 |
Location: |
Chr.16:10545339-10744878 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec16a | NM_001204229.1 | 7 | 1072 | NP_001191158.1 |
NM_177562.5 | 8 | 1078 | NP_808230.2 | |
AK089327.1 | 7 | 1002 | ||
AK122255.1 | 8 | 1030 | BAC65537.1 | |
AK155314.1 | 7 | 1014 | BAE33184.1 | |
AK156927.1 | 7 | 1003 | BAE33899.1 | |
AK170754.1 | 7 | 1003 | BAE42005.1 | |
AK171134.1 | 7 | 1003 | ||
BC138907.1 | 8 | 981 | ||
BC138909.1 | 8 | 981 |