Assay Details
Target Gene Details
Entrez Gene ID: | 320712 |
Gene Name: | ABI gene family, member 3 (NESH) binding protein |
Gene Aliases: |
5033411B22Rik, AI506287, BG172926, D930038M13Rik, TARSH, eratin |
Location: |
Chr.16:56477801-56690135 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abi3bp | NM_001014399.2 | NP_001014421.2 | ||
NM_001014422.2 | NP_001014422.1 | |||
NM_001014423.2 | NP_001014423.1 | |||
NM_001014424.2 | NP_001014424.1 | |||
NM_178790.4 | NP_848905.2 | |||
AB190339.1 | BAD91388.1 | |||
AB190340.1 | BAD91389.1 | |||
AB190341.1 | BAD91390.1 | |||
AB190342.1 | BAD91391.1 | |||
AB190343.1 | BAD91392.1 | |||
AK086582.1 | BAC39696.1 | |||
BC145388.1 | ||||
BC150763.1 |