Assay Details
Target Gene Details
Entrez Gene ID: | 223337 |
Gene Name: | UDP glycosyltransferases 3 family, polypeptide A2 |
Gene Aliases: |
AI313915 |
Location: |
Chr.15:9335551-9370955 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt3a2 | NM_144845.3 | NP_659094.1 | ||
AK041045.1 | BAC30796.1 | |||
AK050128.1 | BAC34080.1 | |||
AK052644.1 | ||||
AK143815.1 | BAE25548.1 | |||
BC022134.1 | AAH22134.1 | |||
BC034837.1 | AAH34837.1 |