Assay Details
Target Gene Details
Entrez Gene ID: | 224273 |
Gene Name: | beta-gamma crystallin domain containing 3 |
Gene Aliases: |
5031404N19, AU022790 |
Location: |
Chr.16:59490775-59601399 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crybg3 | NM_174848.3 | 5 | 2032 | NP_777273.2 |
BC043118.1 | 5 | 2032 | AAH43118.1 |