Assay Details
Target Gene Details
Entrez Gene ID: | 70356 |
Gene Name: | suppression of tumorigenicity 13 |
Gene Aliases: |
1110007I03Rik, 3110002K08Rik, AW555194, HIP, HOP, HSPABP, HSPABP1, PRO0786, SNC6, p48 |
Location: |
Chr.15:81365044-81399694 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
St13 | NM_133726.2 | 11 | 1022 | NP_598487.1 |
AK013981.1 | 11 | 1005 | ||
AK146528.1 | 11 | 1018 | BAE27237.1 | |
AK152270.1 | 11 | 1018 | BAE31086.1 | |
AK168557.1 | 6 | 544 | BAE40430.1 | |
AK205828.1 | 2 | 104 | ||
AK206977.1 | 2 | 104 | ||
BC003843.1 | 11 | 1022 | AAH03843.1 | |
CT010318.1 | 11 | 922 | CAJ18526.1 |