Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mov10l1 | NM_031260.2 | 21 | 2939 | NP_112550.2 |
AF285587.1 | 20 | 2828 | AAK31966.1 | |
AF340211.1 | 7 | 907 | AAK77049.1 | |
AK084786.1 | 5 | 509 | BAC39279.1 | |
AY303754.1 | 5 | 577 | AAP60176.1 | |
JF319145.1 | 21 | 2968 |