Assay Details
Target Gene Details
Entrez Gene ID: | 28075 |
Gene Name: | desumoylating isopeptidase 1 |
Gene Aliases: |
AI427858, AI850401, D15Wsu75e, DeSI-1, Fam152b, Pppde2 |
Location: |
Chr.15:81992523-82016221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Desi1 | NM_134095.2 | NP_598856.1 | ||
AK005706.1 | BAB24198.1 | |||
AK005963.1 | BAB24342.1 | |||
AK016347.1 | ||||
AK131703.1 | BAE20768.1 | |||
AK146155.1 | BAE26939.1 | |||
AK170731.1 | BAE41987.1 | |||
AK189878.1 | ||||
BC022097.1 | AAH22097.1 | |||
BC056972.1 | AAH56972.1 | |||
BC071205.1 | AAH71205.1 | |||
BC131979.1 | ||||
BC132642.1 |
Target Gene Details
Entrez Gene ID: | 14375 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 6 |
Gene Aliases: |
70kDa, G22p1, Ku70 |
Location: |
Chr.15:81987826-82040085 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xrcc6 |
More Information
Set Membership: |
Intragenic Non-exonic |