Assay Details
Target Gene Details
Entrez Gene ID: | 79456 |
Gene Name: | RecQ protein-like 4 |
Gene Aliases: |
RecQ4 |
Location: |
Chr.15:76703553-76710576 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Recql4 | NM_058214.3 | NP_478121.2 | ||
AB039882.1 | BAD11131.1 | |||
AB175741.1 | BAD14289.1 | |||
AK090260.1 | ||||
BC131656.1 |
More Information
Set Membership: |
Intragenic Non-exonic |