Assay Details
Target Gene Details
Entrez Gene ID: | 213053 |
Gene Name: | solute carrier family 39 (zinc transporter), member 14 |
Gene Aliases: |
FAD-123, ZIP-14, Zip14, fad123 |
Location: |
Chr.14:70303467-70351459 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc39a14 | NM_001135151.1 | NP_001128623.1 | ||
NM_001135152.1 | NP_001128624.1 | |||
NM_144808.4 | NP_659057.2 | |||
AB177995.1 | BAD16742.1 | |||
AK090193.1 | ||||
AK122197.1 | BAC65479.2 | |||
BC021530.1 | AAH21530.1 | |||
CB521415.1 |
More Information
Set Membership: |
Intragenic Non-exonic |