Assay Details
Target Gene Details
Entrez Gene ID: | 68966 |
Gene Name: | neuroguidin, EIF4E binding protein |
Gene Aliases: |
1500001L15Rik, Ngd, neuroguidin |
Location: |
Chr.14:55015454-55024138 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ngdn | NM_026890.2 | 4 | 328 | NP_081166.1 |
AK005098.1 | 4 | 210 | BAB23816.1 | |
AK200275.1 | 2 | 78 | ||
AK212411.1 | 2 | 78 | ||
AK214945.1 | 2 | 78 | ||
BC023770.1 | 3 | 213 | AAH23770.1 | |
BC052790.1 | 4 | 266 | AAH52790.1 | |
CN715203.1 | ||||
EF437387.1 |
More Information
Set Membership: |
Intragenic Exonic |