Assay Details
Target Gene Details
Entrez Gene ID: | 218885 |
Gene Name: | oxidoreductase NAD-binding domain containing 1 |
Gene Aliases: |
2410002F01Rik, AW492292, BC019806 |
Location: |
Chr.14:32085728-32103203 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oxnad1 | NM_145460.2 | 6 | 882 | NP_663435.2 |
AK028649.1 | 5 | 3552 | ||
AK031978.1 | 5 | 511 | BAC27636.1 | |
AK032160.1 | ||||
AK075924.1 | 5 | 508 | ||
AK139884.1 | 6 | 882 | BAE24171.1 | |
BC019806.1 | 6 | 660 | AAH19806.1 |
More Information
Set Membership: |
Intragenic Exonic |