Assay Details
Target Gene Details
Entrez Gene ID: | 219151 |
Gene Name: | scavenger receptor class A, member 3 |
Gene Aliases: |
APC7, C130058N24Rik, CSR, CSR1, MSLR1, MSRL1 |
Location: |
Chr.14:65919395-65953744 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scara3 | NM_172604.3 | 1 | 84 | NP_766192.1 |
AK048423.1 | 1 | 85 | BAC33332.1 | |
AK054188.1 | 1 | 82 | BAC35687.1 |
More Information
Set Membership: |
Intragenic Exonic |