Assay Details
Target Gene Details
Entrez Gene ID: | 27374 |
Gene Name: | protein arginine N-methyltransferase 5 |
Gene Aliases: |
Jbp1, Skb1 |
Location: |
Chr.14:54507182-54517470 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prmt5 | NM_013768.3 | 14 | 1647 | NP_038796.2 |
AF167573.1 | 14 | 1515 | AAF04503.1 | |
AK076346.1 | 14 | 1538 | ||
AK132414.1 | 14 | 1539 | BAE21155.1 | |
AK165165.1 | 14 | 1565 | BAE38057.1 | |
AK181096.1 | 3 | 257 | ||
BC023905.1 | 14 | 1594 | AAH23905.1 |
More Information
Set Membership: |
Intragenic Exonic |