Assay Details
Target Gene Details
Entrez Gene ID: | 114741 |
Gene Name: | suppressor of Ty 16 |
Gene Aliases: |
Cdc68, Fact140, Spt16, Supt16h |
Location: |
Chr.14:52160419-52197239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supt16 | NM_033618.3 | NP_291096.2 | ||
AF323667.1 | AAL04452.1 | |||
AK146529.1 | ||||
AK148506.1 | BAE28591.1 | |||
AK169561.1 |
More Information
Set Membership: |
Intragenic Non-exonic |