Assay Details
Target Gene Details
Entrez Gene ID: | 54342 |
Gene Name: | glucosamine-phosphate N-acetyltransferase 1 |
Gene Aliases: |
AU017428, AU040593, EMeg32, Gpnat1, Gsnpat |
Location: |
Chr.14:45376421-45388956 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnpnat1 | NM_019425.2 | NP_062298.1 | ||
AJ001006.1 | CAA04463.1 | |||
AK002466.1 | BAB22120.1 | |||
AK007647.1 | BAB25161.1 | |||
AK007722.1 | BAB25212.1 | |||
AK007764.1 | BAB25240.1 | |||
AK008566.1 | BAB25749.1 | |||
AK011098.1 | BAB27395.1 | |||
AK018499.1 | BAB31241.1 | |||
AK028693.1 | ||||
AK050266.1 | ||||
AK167870.1 | BAE39886.1 | |||
AK168074.1 | BAE40048.1 | |||
BC031116.1 | AAH31116.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |