Assay Details
Target Gene Details
Entrez Gene ID: | 13003 |
Gene Name: | versican |
Gene Aliases: |
5430420N07Rik, 9430051N09, Cspg2, DPEAAE, NG2, PG-M, PG-M(V0), PG-M(V1), hdf |
Location: |
Chr.13:89655309-89742694 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vcan | NM_001081249.1 | 4 | 943 | NP_001074718.1 |
NM_001134474.1 | 4 | 943 | NP_001127946.1 | |
NM_001134475.1 | 4 | 943 | NP_001127947.1 | |
NM_019389.2 | 4 | 943 | NP_062262.2 | |
NM_172955.1 | 4 | 943 | NP_766543.1 | |
AK014525.1 | 4 | 753 | BAB29411.3 | |
AK034871.1 | 4 | 750 | BAC28862.1 | |
AK040503.1 | 1 | 77 | BAC30610.1 | |
AK135324.1 | 4 | 940 | BAE22489.1 | |
AK163927.1 | 4 | 800 | BAE37538.1 | |
AK165069.1 | 4 | 746 | BAE38022.1 | |
BC021652.1 | 4 | 783 | ||
BC096495.1 | 4 | 753 | AAH96495.1 | |
D16263.1 | 3 | 503 | BAA03796.1 | |
D28599.1 | 4 | 675 | ||
D32040.1 | 4 | 675 | BAA06802.1 |
More Information
Set Membership: |
Intragenic Exonic |