Assay Details
Target Gene Details
Entrez Gene ID: | 20499 |
Gene Name: | solute carrier family 12, member 7 |
Gene Aliases: |
AA408796, D13Ertd261e, Kcc4 |
Location: |
Chr.13:73733053-73816755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc12a7 | NM_011390.2 | NP_035520.1 | ||
AF087436.1 | AAD38328.1 | |||
AK052910.1 | BAC35198.1 | |||
AK083598.1 | ||||
AK131129.1 | BAD21379.1 | |||
AK143535.1 | ||||
AK149750.1 | BAE29061.1 | |||
AK166215.1 | ||||
AK169950.1 | BAE41477.1 | |||
AK171498.1 | BAE42491.1 | |||
BC141107.1 |
More Information
Set Membership: |
Intragenic Non-exonic |