Assay Details
Target Gene Details
Entrez Gene ID: | 20499 |
Gene Name: | solute carrier family 12, member 7 |
Gene Aliases: |
AA408796, D13Ertd261e, Kcc4 |
Location: |
Chr.13:73733053-73816755 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc12a7 | NM_011390.2 | 22 | 2962 | NP_035520.1 |
AF087436.1 | 22 | 2964 | AAD38328.1 | |
AK131129.1 | 22 | 2916 | BAD21379.1 | |
AK143535.1 | 22 | 2956 | ||
AK149750.1 | 22 | 2957 | BAE29061.1 | |
AK166215.1 | 22 | 2956 | ||
AK169950.1 | 22 | 2952 | BAE41477.1 | |
BC141107.1 | 22 | 2922 |
More Information
Set Membership: |
Intragenic Exonic |