Assay Details
Target Gene Details
Entrez Gene ID: | 20499 |
Gene Name: | solute carrier family 12, member 7 |
Gene Aliases: |
AA408796, D13Ertd261e, Kcc4 |
Location: |
Chr.13:73733053-73816755 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc12a7 | NM_011390.2 | 2 | 240 | NP_035520.1 |
AF087436.1 | 2 | 242 | AAD38328.1 | |
AK052910.1 | 2 | 291 | BAC35198.1 | |
AK083598.1 | 2 | 198 | ||
AK131129.1 | 2 | 194 | BAD21379.1 | |
AK143535.1 | 2 | 234 | ||
AK149750.1 | 2 | 235 | BAE29061.1 | |
AK166215.1 | 2 | 234 | ||
AK169950.1 | 2 | 230 | BAE41477.1 | |
AK171498.1 | 2 | 277 | BAE42491.1 | |
BC141107.1 | 2 | 200 |
More Information
Set Membership: |
Intragenic Exonic |