Assay Details
Target Gene Details
Entrez Gene ID: | 74338 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19 |
Gene Aliases: |
4632401C08Rik, B<0>AT1 |
Location: |
Chr.13:73679745-73700695 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a19 | NM_028878.3 | 9 | 1295 | NP_083154.1 |
AB096860.1 | 7 | 978 | BAC66209.1 | |
AJ633679.1 | 9 | 1304 | CAG17898.1 | |
AK014544.1 | 9 | 1296 | BAB29422.1 | |
AK028784.1 | 9 | 1296 | BAC26119.1 | |
AK128923.1 | 8 | 1638 | ||
AK162399.1 | 8 | 1040 | BAE36894.1 | |
BC107411.1 | 9 | 1296 | AAI07412.1 | |
BC107412.2 | 9 | 1300 | AAI07413.1 |
More Information
Set Membership: |
Intragenic Exonic |