Assay Details
Target Gene Details
Entrez Gene ID: | 74338 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19 |
Gene Aliases: |
4632401C08Rik, B<0>AT1 |
Location: |
Chr.13:73679745-73700695 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a19 | NM_028878.3 | NP_083154.1 | ||
AB096860.1 | BAC66209.1 | |||
AJ633679.1 | CAG17898.1 | |||
AK014544.1 | BAB29422.1 | |||
AK028784.1 | BAC26119.1 | |||
AK128923.1 | ||||
AK162399.1 | BAE36894.1 | |||
BC107411.1 | AAI07412.1 | |||
BC107412.2 | AAI07413.1 |
More Information
Set Membership: |
Intragenic Non-exonic |