Assay Details
Target Gene Details
Entrez Gene ID: | 22598 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 18 |
Gene Aliases: |
B0AT3, D630001K16Rik, Xt2, Xtrp2 |
Location: |
Chr.13:73661750-73678023 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a18 | NM_001040692.3 | NP_001035782.1 | ||
NM_001136087.2 | NP_001129559.1 | |||
NM_001168643.1 | NP_001162114.1 | |||
NM_001168644.1 | NP_001162115.1 | |||
NM_001168645.1 | NP_001162116.1 | |||
NM_001168646.1 | NP_001162117.1 | |||
AF075262.1 | AAC27757.1 | |||
AF075263.1 | AAC27758.1 | |||
AF075264.1 | AAC27759.1 | |||
AF075265.1 | AAC27760.1 | |||
AF075266.1 | AAC27761.1 | |||
AF075267.1 | AAC27762.1 | |||
AK085258.1 | ||||
AK085473.1 | ||||
AK128907.1 | ||||
BC010748.1 | AAH10748.1 | |||
CA491609.1 |
More Information
Set Membership: |
Intragenic Non-exonic |