Assay Details
Target Gene Details
Entrez Gene ID: | 15387 |
Gene Name: | heterogeneous nuclear ribonucleoprotein K |
Gene Aliases: |
Hnrpk, KBBP, NOVA |
Location: |
Chr.13:58391132-58403343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpk | NM_001301340.1 | NP_001288269.1 | ||
NM_001301341.1 | NP_001288270.1 | |||
NM_001301343.1 | NP_001288272.1 | |||
NM_001301344.1 | NP_001288273.1 | |||
NM_001301345.1 | NP_001288274.1 | |||
NM_025279.3 | NP_079555.1 | |||
AK011428.1 | BAB27614.1 | |||
AK028023.1 | BAC25705.1 | |||
AK051313.1 | BAC34601.1 | |||
AK078777.1 | BAC37389.1 | |||
AK088462.1 | BAC40368.1 | |||
AK128891.1 | ||||
AK135519.1 | BAE22563.1 | |||
AK150848.1 | BAE29905.1 | |||
AK151689.1 | BAE30613.1 | |||
AK152927.1 | BAE31602.1 | |||
AK160615.1 | BAE35915.1 | |||
AK160887.1 | BAE36070.1 | |||
AK166656.1 | BAE38921.1 | |||
AK166927.1 | BAE39121.1 | |||
AK167600.1 | BAE39657.1 | |||
AK169064.1 | BAE40851.1 | |||
BC006694.1 | AAH06694.1 | |||
BC089328.1 | AAH89328.1 | |||
BY794631.1 | ||||
EU604547.1 |
More Information
Set Membership: |
Intragenic Non-exonic |