Assay Details
Target Gene Details
Entrez Gene ID: | 18570 |
Gene Name: | programmed cell death 6 |
Gene Aliases: |
AV299538, Alg2, MA-3, PS2, alg-2 |
Location: |
Chr.13:74303121-74317575 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdcd6 | NM_011051.3 | NP_035181.1 | ||
AK008610.1 | BAB25775.1 | |||
AK028223.1 | BAC25823.1 | |||
AK078044.1 | BAC37107.1 | |||
AK147979.1 | BAE28264.1 | |||
AK150335.1 | BAE29477.1 | |||
BC040079.1 | AAH40079.1 | |||
BU604816.1 | ||||
U49112.1 | AAB38108.1 | |||
X14938.1 | CAA33064.1 |
More Information
Set Membership: |
Intragenic Non-exonic |