Assay Details
Target Gene Details
Entrez Gene ID: | 66143 |
Gene Name: | eukaryotic translation elongation factor 1 epsilon 1 |
Gene Aliases: |
1110003A02Rik |
Location: |
Chr.13:38645691-38659028 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eef1e1 | NM_025380.2 | NP_079656.1 | ||
AK003335.1 | BAB22723.1 | |||
AK009889.1 | ||||
AK199512.1 | ||||
BC048466.1 | AAH48466.1 | |||
EF437326.1 |
More Information
Set Membership: |
Intragenic Non-exonic |