Assay Details
Target Gene Details
Entrez Gene ID: | 328059 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 15 |
Gene Aliases: |
Arpat |
Location: |
Chr.12:8528481-8599066 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a15 | NM_001038660.2 | 2 | 853 | NP_001033749.1 |
NM_177802.3 | 3 | 410 | NP_808470.1 | |
AK033481.1 | 3 | 410 | BAC28311.1 | |
AY795939.1 | 2 | 832 | AAX09981.1 |
More Information
Set Membership: |
Intragenic Exonic |