Assay Details
Target Gene Details
Entrez Gene ID: | 208439 |
Gene Name: | kelch-like 29 |
Gene Aliases: |
A230106N14Rik, Gm68, Kbtbd9, mKIAA1921 |
Location: |
Chr.12:5077465-5375682 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhl29 | NM_001164493.1 | 6 | 1748 | NP_001157965.1 |
AK122572.1 | 6 | 1401 | BAC65854.1 | |
BC138283.1 | 5 | 1104 | ||
BC145230.1 | 5 | 1104 | ||
BC145748.1 | 5 | 1104 |
More Information
Set Membership: |
Intragenic Exonic |