Assay Details
Target Gene Details
Entrez Gene ID: | 18035 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha |
Gene Aliases: |
AI462015, Nfkbi |
Location: |
Chr.12:55489409-55492647 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfkbia | NM_010907.2 | 3 | 529 | NP_035037.2 |
AK089215.1 | 3 | 478 | ||
AK151115.1 | 3 | 482 | BAE30124.1 | |
AK151608.1 | 3 | 482 | BAE30547.1 | |
AK161622.1 | 3 | 441 | ||
BC046754.1 | 3 | 462 | AAH46754.1 | |
U36277.1 | 3 | 473 | AAA79696.1 |