Assay Details
Target Gene Details
Entrez Gene ID: | 18035 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha |
Gene Aliases: |
AI462015, Nfkbi |
Location: |
Chr.12:55489409-55492647 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfkbia | NM_010907.2 | NP_035037.2 | ||
AK089215.1 | ||||
AK151115.1 | BAE30124.1 | |||
AK151608.1 | BAE30547.1 | |||
AK161622.1 | ||||
BC046754.1 | AAH46754.1 | |||
U36277.1 | AAA79696.1 |