Assay Details
Target Gene Details
Entrez Gene ID: | 27277 |
Gene Name: | golgi autoantigen, golgin subfamily a, 5 |
Gene Aliases: |
Ret-II |
Location: |
Chr.12:102469134-102497911 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golga5 | NM_001199004.1 | NP_001185933.1 | ||
NM_013747.4 | NP_038775.1 | |||
AB016784.1 | BAA33010.1 | |||
AF026274.1 | AAF21628.1 | |||
AK007592.1 | ||||
AK138455.1 | BAE23668.1 | |||
AK152533.1 | BAE31289.1 | |||
AK153010.1 | BAE31649.1 | |||
AK168765.1 | BAE40601.1 | |||
BC016883.1 | AAH16883.1 | |||
BC086782.1 | AAH86782.1 |