Assay Details
Target Gene Details
Entrez Gene ID: | 104806 |
Gene Name: | Fanconi anemia, complementation group M |
Gene Aliases: |
AI427100, C730036B14Rik, D12Ertd364e |
Location: |
Chr.12:65074091-65132058 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fancm | NM_178912.3 | NP_849243.2 | ||
AK039944.1 | BAC30483.1 | |||
AK050306.1 | BAC34178.2 | |||
AK053715.1 | BAC35487.1 | |||
AK084697.1 | BAC39257.2 | |||
AK086395.1 | BAC39660.1 | |||
AK173211.1 | BAD32489.1 | |||
BC150785.1 |
More Information
Set Membership: |
Intragenic Non-exonic |