Assay Details
Target Gene Details
Entrez Gene ID: | 52504 |
Gene Name: | centromere protein O |
Gene Aliases: |
2810429O05Rik, 8430427C03Rik, D12Ertd482e |
Location: |
Chr.12:4197058-4234294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpo | NM_134046.5 | NP_598807.2 | ||
AK013198.1 | BAB28707.1 | |||
AK018444.1 | ||||
AK029226.1 | BAC26354.1 | |||
AK036089.1 | BAC29301.1 | |||
AK134266.1 | BAE22073.1 | |||
BC023148.1 | AAH23148.1 | |||
BC034399.1 | AAH34399.1 |