Assay Details
Target Gene Details
Entrez Gene ID: | 76547 |
Gene Name: | transmembrane protein 101 |
Gene Aliases: |
2610511E22Rik |
Location: |
Chr.11:102152547-102156407 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem101 | NM_029649.2 | 3 | 458 | NP_083925.2 |
AK012106.1 | 3 | 412 | BAB28037.1 | |
AK041154.1 | 3 | 417 | BAC30841.1 | |
AK049378.1 | 3 | 440 | BAC33721.1 | |
AK140260.1 | 3 | 459 | BAE24302.1 | |
BC011109.1 | 3 | 428 | AAH11109.1 |