Assay Details
Target Gene Details
Entrez Gene ID: | 327978 |
Gene Name: | schlafen 5 |
Gene Aliases: |
AI956840 |
Location: |
Chr.11:82951349-82964850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slfn5 | NM_183201.4 | NP_899024.3 | ||
AK036486.1 | BAC29447.1 | |||
AK040359.1 | BAE20578.1 | |||
AK171886.1 | BAE42719.1 | |||
AY261804.1 | AAP30074.1 | |||
AY261805.1 | AAP30075.1 | |||
BC117909.1 | ||||
BC117910.1 |
Target Gene Details
Entrez Gene ID: | 76392 |
Gene Name: | schlafen 5, opposite strand |
Gene Aliases: |
1700003F17Rik |
Location: |
Chr.11:82942341-82960681 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slfn5os | NR_045932.1 | |||
AK005634.1 | BAB24160.1 | |||
BY705829.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |