Assay Details
Target Gene Details
Entrez Gene ID: | 19184 |
Gene Name: | protease (prosome, macropain) 26S subunit, ATPase 5 |
Gene Aliases: |
mSUG1 |
Location: |
Chr.11:106256185-106263112 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmc5 | NM_008950.1 | NP_032976.1 | ||
AK010505.1 | BAB26990.1 | |||
AK012294.1 | ||||
AK135451.1 | BAE22539.1 | |||
AK145703.1 | BAE26599.1 | |||
BC030840.1 | AAH30840.1 | |||
Z54219.1 | CAA90961.1 |
More Information
Set Membership: |
Intragenic Non-exonic |