Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc43a2 | NM_001199283.1 | NP_001186212.1 | ||
NM_001199284.1 | NP_001186213.1 | |||
NM_173388.2 | NP_775564.1 | |||
AB120363.1 | BAD91090.1 | |||
AK143773.1 | BAE25533.1 | |||
AK158440.1 | BAE34511.1 | |||
AK159144.1 | BAE34852.1 | |||
AK171104.1 | ||||
AK172685.1 | ||||
BC042513.1 | AAH42513.1 |