Assay Details
Target Gene Details
Entrez Gene ID: | 69806 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 11 |
Gene Aliases: |
1810074D23Rik |
Location: |
Chr.11:113244851-113650752 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc39a11 | NM_001166503.1 | NP_001159975.1 | ||
NM_027216.5 | NP_081492.4 | |||
AK007986.1 | BAB25390.1 | |||
AK036999.1 | BAC29664.1 | |||
AK049368.1 | BAC33713.1 | |||
AK165545.1 | ||||
AK170673.1 | BAE41950.1 | |||
AK192534.1 | ||||
BC019647.1 | AAH19647.1 | |||
BY299357.1 |
More Information
Set Membership: |
Intragenic Non-exonic |