Assay Details
Target Gene Details
Entrez Gene ID: | 75273 |
Gene Name: | proline, glutamic acid and leucine rich protein 1 |
Gene Aliases: |
4930563C04Rik, MNAR |
Location: |
Chr.11:70392881-70410031 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pelp1 | NM_029231.4 | NP_083507.3 | ||
AK005027.1 | BAB23754.1 | |||
AK128966.1 | ||||
AK211280.1 | ||||
BC016444.1 | AAH16444.1 | |||
BC057987.1 | AAH57987.1 | |||
BC090620.1 |