Assay Details
Target Gene Details
Entrez Gene ID: | 75273 |
Gene Name: | proline, glutamic acid and leucine rich protein 1 |
Gene Aliases: |
4930563C04Rik, MNAR |
Location: |
Chr.11:70392881-70410031 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pelp1 | NM_029231.4 | 13 | 1529 | NP_083507.3 |
AK005027.1 | 13 | 1527 | BAB23754.1 | |
AK128966.1 | 13 | 1529 | ||
BC016444.1 | 3 | 346 | AAH16444.1 | |
BC057987.1 | 7 | 757 | AAH57987.1 | |
BC090620.1 | 13 | 1523 |