Assay Details
Target Gene Details
Entrez Gene ID: | 20415 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP |
Location: |
Chr.11:69610281-69618025 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shbg | NM_011367.2 | 7 | 1011 | NP_035497.1 |
AK010959.1 | 7 | 1010 | ||
BC120785.1 | 7 | 996 | ||
BC120787.1 | 7 | 996 | ||
U85644.1 | 7 | 974 | AAB42088.1 |