Assay Details
Target Gene Details
Entrez Gene ID: | 20415 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP |
Location: |
Chr.11:69610281-69618025 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shbg | NM_011367.2 | NP_035497.1 | ||
AK010959.1 | ||||
BC120785.1 | ||||
BC120787.1 | ||||
U85644.1 | AAB42088.1 |