Assay Details
Target Gene Details
Entrez Gene ID: | 66397 |
Gene Name: | SAR1 gene homolog B (S. cerevisiae) |
Gene Aliases: |
2310075M17Rik, 2900019I22Rik, CMRD, Sara1b, Sara2, Sarb |
Location: |
Chr.11:51763663-51791953 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sar1b | NM_025535.2 | 5 | 444 | NP_079811.1 |
AK002327.1 | 5 | 421 | BAB22015.1 | |
AK010187.1 | 5 | 427 | BAB26755.1 | |
AK013559.1 | 5 | 406 | BAB28905.1 | |
AK150906.1 | 5 | 400 | BAE29948.1 | |
AK155429.1 | 5 | 414 | BAE33261.1 | |
BC082550.1 | 5 | 413 | AAH82550.1 | |
BC085178.1 | 5 | 397 |