Assay Details
Target Gene Details
Entrez Gene ID: | 70357 |
Gene Name: | Kv channel-interacting protein 1 |
Gene Aliases: |
KCHIP1, Kchip1.2 |
Location: |
Chr.11:33629339-33993193 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnip1 | NM_001190886.1 | NP_001177815.1 | ||
AK048178.1 | ||||
AK081845.1 | BAC38347.1 | |||
AK082267.1 | ||||
DQ148494.1 | AAZ77811.1 |
Target Gene Details
Entrez Gene ID: | 16533 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 1 |
Gene Aliases: |
BKbeta1 |
Location: |
Chr.11:33962951-33973641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnmb1 | NM_031169.4 | NP_112446.2 | ||
AF020711.1 | AAD11857.1 | |||
AJ001291.1 | CAA04651.1 | |||
AK038987.1 | BAC30193.1 | |||
BC013338.1 | AAH13338.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |